Assay Details
Target Gene Details
Entrez Gene ID: | 8470 |
Gene Name: | sorbin and SH3 domain containing 2 |
Gene Aliases: |
ARGBP2, PRO0618 |
Location: |
Chr.4:185585444-185956716 on Build GRCh38 |
Assay Gene Location: | Within Exon 34 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1020731 | Chr.4:185591279 - 185802877 on Build GRCh38 | Gain |
|
| nsv1028146 | Chr.4:185582745 - 186417181 on Build GRCh38 | Gain |
|
| nsv527026 | Chr.4:185404275 - 185632326 on Build GRCh38 | Gain |
|
| nsv830169 | Chr.4:185533910 - 185717009 on Build GRCh38 | Gain+Loss |
|
| esv3603509 | Chr.4:185579224 - 185611747 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map