Assay Details
Target Gene Details
Entrez Gene ID: | 152789 |
Gene Name: | janus kinase and microtubule interacting protein 1 |
Gene Aliases: |
Gababrbp, JAMIP1, MARLIN1 |
Location: |
Chr.4:6024925-6200591 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JAKMIP1 | NM_001099433.1 | NP_001092903.1 | ||
| XM_017007788.1 | XP_016863277.1 | |||
| XM_017007789.1 | XP_016863278.1 | |||
| XM_017007790.1 | XP_016863279.1 | |||
| XM_017007791.1 | XP_016863280.1 | |||
| XM_017007792.1 | XP_016863281.1 | |||
| XM_017007793.1 | XP_016863282.1 | |||
| XM_017007794.1 | XP_016863283.1 | |||
| XM_017007795.1 | XP_016863284.1 | |||
| XM_017007797.1 | XP_016863286.1 | |||
| XM_017007798.1 | XP_016863287.1 | |||
| XM_017007799.1 | XP_016863288.1 | |||
| AM412307.1 | ||||
| AM412308.1 | ||||
| AM412309.1 | ||||
| AM412310.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4209 | Chr.4:6018928 - 6052309 on Build GRCh38 | Insertion |
|
| nsv461185 | Chr.4:6041005 - 6063703 on Build GRCh38 | Loss |
|
| nsv829845 | Chr.4:5870068 - 6088797 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map