Assay Details
Target Gene Details
Entrez Gene ID: | 101930370 |
Gene Name: | uncharacterized LOC101930370 |
Gene Aliases: |
- |
Location: |
Chr.4:173131933-173169652 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101930370 | NR_134241.1 | 4 | 2111 | |
| NR_134242.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3603208 | Chr.4:173140942 - 173145440 on Build GRCh38 | Loss |
|

Genomic Map