Assay Details
Target Gene Details
Entrez Gene ID: | 23173 |
Gene Name: | methionyl aminopeptidase 1 |
Gene Aliases: |
MAP1A, MetAP1A |
Location: |
Chr.4:98995637-99062809 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| METAP1 | NM_015143.2 | NP_055958.2 | ||
| XM_011531779.1 | XP_011530081.1 | |||
| XM_017007914.1 | XP_016863403.1 | |||
| XM_017007915.1 | XP_016863404.1 | |||
| AK304239.1 | ||||
| D42084.1 | ||||
| DA178462.1 |
More Information
Set Membership: |
|

Genomic Map