Assay Details
Target Gene Details
Entrez Gene ID: | 886 |
Gene Name: | cholecystokinin A receptor |
Gene Aliases: |
CCK-A, CCK1-R, CCK1R, CCKRA |
Location: |
Chr.4:26481396-26490420 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCKAR | NM_000730.2 | NP_000721.1 | ||
| AK313978.1 | ||||
| AY322549.1 | AAP84362.1 | |||
| BC074987.2 | AAH74987.1 | |||
| L13605.1 | AAA35659.1 | |||
| L19315.1 | AAA02819.1 |
More Information
Set Membership: |
|

Genomic Map