Assay Details
Target Gene Details
Entrez Gene ID: | 22902 |
Gene Name: | RUN and FYVE domain containing 3 |
Gene Aliases: |
RIPX, SINGAR1, ZFYVE30 |
Location: |
Chr.4:70704204-70808622 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RUFY3 | NM_001130709.1 | NP_001124181.1 | ||
| XM_011531750.2 | XP_011530052.1 | |||
| XM_011531751.2 | XP_011530053.1 | |||
| XM_011531752.2 | XP_011530054.1 | |||
| XM_011531753.2 | XP_011530055.1 | |||
| XM_011531754.2 | XP_011530056.1 | |||
| XM_011531755.2 | XP_011530057.1 | |||
| XM_011531756.2 | XP_011530058.1 | |||
| XM_017007890.1 | XP_016863379.1 | |||
| AF112221.1 | AAF17208.1 | |||
| AK000911.1 | ||||
| BM920380.1 | ||||
| DB483639.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829965 | Chr.4:70630001 - 70806591 on Build GRCh38 | Gain |
|
| nsv1012495 | Chr.4:69979953 - 70947088 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72861330,rs77206902] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map