Assay Details
Target Gene Details
Entrez Gene ID: | 8603 |
Gene Name: | family with sequence similarity 193 member A |
Gene Aliases: |
C4orf8, RES4-22 |
Location: |
Chr.4:2596288-2732575 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM193A | NM_001256666.1 | NP_001243595.1 | ||
| NM_001256667.1 | NP_001243596.1 | |||
| NM_001256668.1 | NP_001243597.1 | |||
| NM_003704.3 | NP_003695.3 | |||
| NR_046335.1 | ||||
| NR_046336.1 | ||||
| XM_006713930.2 | XP_006713993.2 | |||
| XM_006713932.2 | XP_006713995.2 | |||
| XM_011513590.1 | XP_011511892.1 | |||
| XM_011513593.2 | XP_011511895.1 | |||
| XM_017008788.1 | XP_016864277.1 | |||
| AB000459.1 | BAA19116.1 | |||
| AB000460.1 | BAA19117.1 | |||
| AB000461.1 | BAA19118.1 | |||
| AK308449.1 | ||||
| BC136646.1 | ||||
| BC144340.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829839 | Chr.4:2622695 - 2657883 on Build GRCh38 | Loss |
|
| esv2726835 | Chr.4:2019561 - 2959568 on Build GRCh38 | Deletion |
|

Genomic Map