Assay Details
Target Gene Details
Entrez Gene ID: | 166824 |
Gene Name: | Ras association domain family member 6 |
Gene Aliases: |
- |
Location: |
Chr.4:73571550-73620631 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RASSF6 | NM_001270391.1 | NP_001257320.1 | ||
| NM_001270392.1 | NP_001257321.1 | |||
| NM_177532.4 | NP_803876.1 | |||
| NM_201431.2 | NP_958834.1 | |||
| XM_011531687.2 | XP_011529989.1 | |||
| XM_011531688.1 | XP_011529990.1 | |||
| XM_011531689.2 | XP_011529991.1 | |||
| XM_011531690.2 | XP_011529992.1 | |||
| XM_011531691.1 | XP_011529993.1 | |||
| XM_017007837.1 | XP_016863326.1 | |||
| XM_017007838.1 | 7 | 2602 | XP_016863327.1 | |
| AK126346.1 | BAC86530.1 | |||
| AY217664.1 | AAO61689.1 | |||
| AY217665.1 | AAO61690.1 | |||
| BC058835.1 | AAH58835.1 | |||
| CR749283.1 | CAH18138.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1001725 | Chr.4:73488333 - 73995207 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72863008] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map