Assay Details
Target Gene Details
Entrez Gene ID: | 132864 |
Gene Name: | cytoplasmic polyadenylation element binding protein 2 |
Gene Aliases: |
CPE-BP2, CPEB-2, hCPEB-2 |
Location: |
Chr.4:15002451-15070153 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPEB2 | NM_001177381.1 | NP_001170852.1 | ||
| NM_001177382.1 | NP_001170853.1 | |||
| NM_001177383.1 | NP_001170854.1 | |||
| NM_001177384.1 | NP_001170855.1 | |||
| NM_182485.2 | NP_872291.2 | |||
| NM_182646.2 | NP_872587.2 | |||
| XM_005248135.3 | 1 | 220 | XP_005248192.2 | |
| XM_011513777.2 | 1 | 220 | XP_011512079.1 | |
| XM_011513778.2 | 1 | 220 | XP_011512080.1 | |
| XM_011513779.1 | 1 | 220 | XP_011512081.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829868 | Chr.4:14962935 - 15163082 on Build GRCh38 | Gain |
|
| dgv8976n54 | Chr.4:15002272 - 15005183 on Build GRCh38 | Gain+Loss |
|
| nsv593758 | Chr.4:15001808 - 15003839 on Build GRCh38 | Gain |
|
| dgv8977n54 | Chr.4:15002329 - 15004512 on Build GRCh38 | Loss |
|
| nsv461280 | Chr.4:14960872 - 15079750 on Build GRCh38 | Loss |
|
| esv29275 | Chr.4:15001712 - 15005401 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map