Assay Details
Target Gene Details
Entrez Gene ID: | 22902 |
Gene Name: | RUN and FYVE domain containing 3 |
Gene Aliases: |
RIPX, SINGAR1, ZFYVE30 |
Location: |
Chr.4:70704204-70808622 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829965 | Chr.4:70630001 - 70806591 on Build GRCh38 | Gain |
|
| nsv4380 | Chr.4:70720666 - 70754529 on Build GRCh38 | Insertion |
|
| nsv1012495 | Chr.4:69979953 - 70947088 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map