Assay Details
Target Gene Details
Entrez Gene ID: | 91050 |
Gene Name: | coiled-coil domain containing 149 |
Gene Aliases: |
- |
Location: |
Chr.4:24806117-24980204 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC149 | NM_001130726.2 | 12 | 2567 | NP_001124198.1 |
| NM_173463.4 | 13 | 2668 | NP_775734.2 | |
| XM_005248210.3 | 13 | 2629 | XP_005248267.1 | |
| XM_011513906.2 | 15 | 2711 | XP_011512208.1 | |
| XM_011513907.2 | 14 | 2584 | XP_011512209.1 | |
| XM_011513908.1 | 13 | 2506 | XP_011512210.1 | |
| XM_017008827.1 | 14 | 2678 | XP_016864316.1 | |
| XM_017008828.1 | 13 | 2602 | XP_016864317.1 | |
| AK023472.1 | 1 | 825 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829887 | Chr.4:24771923 - 25000466 on Build GRCh38 | Gain+Loss |
|

Genomic Map