Assay Details
Target Gene Details
Entrez Gene ID: | 132884 |
Gene Name: | EvC ciliary complex subunit 2 |
Gene Aliases: |
LBN, WAD |
Location: |
Chr.4:5531921-5709548 on Build GRCh38 |
Assay Gene Location: | Within Intron 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EVC2 | NM_001166136.1 | NP_001159608.1 | ||
| NM_147127.4 | NP_667338.3 | |||
| XM_017007735.1 | XP_016863224.1 | |||
| XM_017007736.1 | XP_016863225.1 | |||
| XM_017007737.1 | XP_016863226.1 | |||
| XM_017007739.1 | XP_016863228.1 | |||
| AB083067.1 | BAC06590.1 | |||
| AY152402.1 | AAN86577.1 | |||
| AY152403.1 | AAN86578.1 | |||
| AY185210.1 | AAO22066.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012934 | Chr.4:5534522 - 5568113 on Build GRCh38 | Gain |
|
| nsv4207 | Chr.4:5562258 - 5594118 on Build GRCh38 | Insertion |
|
| nsv593480 | Chr.4:4961200 - 5760434 on Build GRCh38 | Loss |
|
| nsv593482 | Chr.4:5184198 - 5593242 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73072291] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map