Assay Details
Target Gene Details
Entrez Gene ID: | 57533 |
Gene Name: | TBC1 domain family member 14 |
Gene Aliases: |
- |
Location: |
Chr.4:6909444-7033118 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBC1D14 | NM_001113361.1 | NP_001106832.1 | ||
| NM_020773.2 | NP_065824.2 | |||
| XM_005247986.2 | XP_005248043.1 | |||
| XM_006713895.2 | XP_006713958.1 | |||
| XM_011513507.2 | XP_011511809.1 | |||
| XM_011513508.1 | XP_011511810.1 | |||
| XM_017008477.1 | XP_016863966.1 | |||
| BC041167.1 | AAH41167.1 | |||
| DA317176.1 | ||||
| DA492919.1 | ||||
| DA754761.1 |
More Information
Set Membership: |
|

Genomic Map