Assay Details
Target Gene Details
Entrez Gene ID: | 10393 |
Gene Name: | anaphase promoting complex subunit 10 |
Gene Aliases: |
APC10, DOC1 |
Location: |
Chr.4:144967117-145098541 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3602553 | Chr.4:145007989 - 145016351 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map