Assay Details
Target Gene Details
Entrez Gene ID: | 646603 |
Gene Name: | chromosome 4 open reading frame 51 |
Gene Aliases: |
- |
Location: |
Chr.4:145680103-145792248 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C4orf51 | XM_017008548.1 | XP_016864037.1 | ||
| XM_017008549.1 | XP_016864038.1 | |||
| XM_017008552.1 | XP_016864041.1 | |||
| XM_017008554.1 | XP_016864043.1 | |||
| XM_017008555.1 | XP_016864044.1 | |||
| XM_017008556.1 | XP_016864045.1 |
Target Gene Details
Entrez Gene ID: | 152485 |
Gene Name: | zinc finger protein 827 |
Gene Aliases: |
- |
Location: |
Chr.4:145757627-145939008 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF827 | NM_001306215.1 | NP_001293144.1 | ||
| NM_178835.4 | NP_849157.2 | |||
| XM_011531634.2 | XP_011529936.1 | |||
| XM_011531635.2 | XP_011529937.1 | |||
| XM_011531645.2 | XP_011529947.1 | |||
| XM_017007768.1 | XP_016863257.1 | |||
| XM_017007769.1 | XP_016863258.1 | |||
| XM_017007770.1 | XP_016863259.1 | |||
| XM_017007771.1 | XP_016863260.1 | |||
| XM_017007772.1 | XP_016863261.1 | |||
| XM_017007773.1 | XP_016863262.1 | |||
| XM_017007774.1 | XP_016863263.1 | |||
| XM_017007775.1 | XP_016863264.1 | |||
| XM_017007777.1 | XP_016863266.1 | |||
| AB074279.1 | ||||
| AF450485.1 | AAP97679.1 | |||
| AK056492.1 | ||||
| AK091130.1 | BAC03591.1 | |||
| AK094136.1 | ||||
| AK225805.1 | ||||
| BC117407.1 | ||||
| BC143577.1 |
More Information
Set Membership: |
|

Genomic Map