Assay Details
Target Gene Details
Entrez Gene ID: | 729830 |
Gene Name: | family with sequence similarity 160 member A1 |
Gene Aliases: |
- |
Location: |
Chr.4:151409143-151670503 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM160A1 | NM_001109977.1 | NP_001103447.1 | ||
| XM_005263199.1 | XP_005263256.1 | |||
| XM_011532221.2 | XP_011530523.1 | |||
| XM_011532222.2 | XP_011530524.1 | |||
| XM_011532223.2 | XP_011530525.1 | |||
| DA722299.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3602685 | Chr.4:151421545 - 151422412 on Build GRCh38 | Loss |
|
| esv3564797 | Chr.4:151421500 - 151422325 on Build GRCh38 | Deletion |
|
| esv2728538 | Chr.4:151421612 - 151422264 on Build GRCh38 | Deletion |
|
| esv2670825 | Chr.4:151421569 - 151422285 on Build GRCh38 | Deletion |
|
| nsv1028814 | Chr.4:150874356 - 152285445 on Build GRCh38 | Gain |
|

Genomic Map