Assay Details
Target Gene Details
Entrez Gene ID: | 6622 |
Gene Name: | synuclein alpha |
Gene Aliases: |
NACP, PARK1, PARK4, PD1 |
Location: |
Chr.4:89724099-89838324 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNCA | NM_000345.3 | 6 | 2711 | NP_000336.1 |
| NM_001146054.1 | 6 | 2707 | NP_001139526.1 | |
| NM_001146055.1 | 6 | 2518 | NP_001139527.1 | |
| NM_007308.2 | 5 | 2623 | NP_009292.1 | |
| JN709859.1 | 5 | 2665 | ||
| JN709860.1 | 4 | 2581 | ||
| JN709861.1 | 5 | 2627 | ||
| JN709862.1 | 5 | 2669 | ||
| JN709863.1 | 4 | 2585 | ||
| JN709864.1 | 5 | 2434 | ||
| JN709865.1 | 5 | 2476 | ||
| JN709866.1 | 4 | 2392 | ||
| JN709867.1 | 5 | 2476 | ||
| JN709868.1 | 5 | 2518 | ||
| JN709869.1 | 4 | 2434 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012406 | Chr.4:89246630 - 90245636 on Build GRCh38 | Loss |
|
| nsv594806 | Chr.4:88621130 - 90539537 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map