Assay Details
Target Gene Details
Entrez Gene ID: | 389197 |
Gene Name: | chromosome 4 open reading frame 50 |
Gene Aliases: |
- |
Location: |
Chr.4:5897591-6018762 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C4orf50 | XM_011513603.2 | XP_011511905.1 | ||
| XM_011513604.2 | XP_011511906.1 | |||
| XM_017008891.1 | XP_016864380.1 | |||
| XM_017008892.1 | XP_016864381.1 | |||
| XM_017008893.1 | XP_016864382.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829844 | Chr.4:5719377 - 5925385 on Build GRCh38 | Gain |
|
| nsv525088 | Chr.4:5917982 - 5924824 on Build GRCh38 | Loss |
|
| nsv829845 | Chr.4:5870068 - 6088797 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs111950929,rs73067428] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map