Assay Details
Target Gene Details
Entrez Gene ID: | 399 |
Gene Name: | ras homolog family member H |
Gene Aliases: |
ARHH, TTF |
Location: |
Chr.4:40191011-40244764 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950578 | Chr.4:40196981 - 40206280 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map