Assay Details
Target Gene Details
Entrez Gene ID: | 84992 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class Y |
Gene Aliases: |
HPMRS6, PIG-Y |
Location: |
Chr.4:88520978-88523801 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIGY | NM_001042616.2 | NP_001036081.1 | ||
| AB206972.1 | BAE44507.1 | |||
| BC007876.2 | ||||
| BI548853.1 |
Target Gene Details
Entrez Gene ID: | 100996939 |
Gene Name: | PIGY upstream reading frame |
Gene Aliases: |
PREY |
Location: |
Chr.4:88520978-88523801 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PYURF | NM_032906.4 | NP_116295.1 | ||
| AK312082.1 | ||||
| BG706896.1 | ||||
| CV812938.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs67364668] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map