Assay Details
Target Gene Details
Entrez Gene ID: | 1482 |
Gene Name: | NK2 homeobox 5 |
Gene Aliases: |
CHNG5, CSX, CSX1, HLHS2, NKX2.5, NKX2E, NKX4-1, VSD3 |
Location: |
Chr.5:173232104-173235312 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NKX2-5 | NM_001166175.1 | 1 | 99 | NP_001159647.1 |
| NM_001166176.1 | 1 | 99 | NP_001159648.1 | |
| NM_004387.3 | 1 | 99 | NP_004378.1 | |
| XM_017009071.1 | XP_016864560.1 | |||
| AK290615.1 | ||||
| AK297844.1 | ||||
| AK307218.1 | ||||
| AK307247.1 | 1 | 51 | ||
| AK307249.1 | ||||
| AK309495.1 | ||||
| BC025711.1 | AAH25711.1 | |||
| DA559427.1 | 1 | 99 | ||
| U34962.1 | 1 | 46 | AAC50470.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950444 | Chr.5:173228498 - 173240197 on Build GRCh38 | Deletion |
|
| esv3575898 | Chr.5:173222243 - 173235416 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map