Assay Details
Target Gene Details
Entrez Gene ID: | 553103 |
Gene Name: | uncharacterized LOC553103 |
Gene Aliases: |
- |
Location: |
Chr.5:132311276-132369916 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC553103 | NR_110997.1 |
Target Gene Details
Entrez Gene ID: | 6583 |
Gene Name: | solute carrier family 22 member 4 |
Gene Aliases: |
OCTN1 |
Location: |
Chr.5:132294384-132344206 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC22A4 | NM_003059.2 | NP_003050.2 | ||
| XM_006714675.3 | XP_006714738.1 | |||
| XM_011543589.2 | XP_011541891.1 | |||
| XM_017009776.1 | XP_016865265.1 | |||
| AB007448.1 | BAA23356.1 | |||
| AK310818.1 | ||||
| BC028313.1 | AAH28313.1 | |||
| Y09881.1 | CAA71007.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs75539158] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map