Assay Details
Target Gene Details
Entrez Gene ID: | 28965 |
Gene Name: | solute carrier family 27 member 6 |
Gene Aliases: |
ACSVL2, FACVL2, FATP6, VLCS-H1 |
Location: |
Chr.5:128965127-129033642 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC27A6 | NM_001017372.2 | 1 | 227 | NP_001017372.1 |
| NM_001317984.1 | 1 | 227 | NP_001304913.1 | |
| NM_014031.4 | 1 | 227 | NP_054750.1 | |
| BC041945.1 | 1 | 226 | AAH41945.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3606738 | Chr.5:128954264 - 128969226 on Build GRCh38 | Loss |
|
| nsv830485 | Chr.5:128962929 - 129144799 on Build GRCh38 | Gain |
|
| esv2656691 | Chr.5:128941074 - 128971417 on Build GRCh38 | Deletion |
|
| nsv819884 | Chr.5:128965366 - 128967561 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map