Assay Details
Target Gene Details
Entrez Gene ID: | 4338 |
Gene Name: | molybdenum cofactor synthesis 2 |
Gene Aliases: |
MCBPE, MOCO1, MOCODB, MPTS |
Location: |
Chr.5:53095679-53109772 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MOCS2 | NM_004531.4 | NP_004522.1 | ||
| NM_176806.3 | NP_789776.1 | |||
| AF091871.1 | AAD14598.1 | |||
| AF117815.1 | AAD13296.1 | |||
| AF155659.1 | AAF67478.1 | |||
| AK126211.1 | ||||
| AK312887.1 | ||||
| BC046097.1 | AAH46097.1 | |||
| CR457172.1 | CAG33453.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830296 | Chr.5:52913698 - 53114423 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75330104,rs76525944] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map