Assay Details
Target Gene Details
Entrez Gene ID: | 1070 |
Gene Name: | centrin 3 |
Gene Aliases: |
CDC31, CEN3 |
Location: |
Chr.5:90393335-90409786 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CETN3 | NM_001297765.1 | NP_001284694.1 | ||
| NM_001297768.1 | NP_001284697.1 | |||
| NM_004365.3 | NP_004356.2 | |||
| BC005383.1 | AAH05383.1 | |||
| BC093793.1 | AAH93793.1 | |||
| BC112040.1 | AAI12041.1 | |||
| BM553126.1 | ||||
| BT006688.1 | AAP35334.1 | |||
| BU569574.1 | ||||
| BU633961.1 | ||||
| CB988525.1 | ||||
| CB998764.1 | ||||
| CR450346.1 | CAG29342.1 | |||
| Y12473.1 | CAA73077.1 |
More Information
Set Membership: |
|

Genomic Map