Assay Details
Target Gene Details
Entrez Gene ID: | 340075 |
Gene Name: | arylsulfatase family member I |
Gene Aliases: |
SPG66 |
Location: |
Chr.5:150296346-150302962 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARSI | NM_001012301.2 | 1 | 213 | NP_001012301.1 |
| AB448735.1 | 1 | 204 | ||
| CN369733.1 | 1 | 112 | ||
| DA264664.1 | ||||
| DA759270.1 | 1 | 213 |
More Information
Set Membership: |
|

Genomic Map