Assay Details
Target Gene Details
Entrez Gene ID: | 5201 |
Gene Name: | prefoldin subunit 1 |
Gene Aliases: |
PDF, PFD1 |
Location: |
Chr.5:140245050-140303141 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PFDN1 | NM_002622.4 | NP_002613.2 | ||
| XM_005268465.3 | XP_005268522.1 | |||
| AK223394.1 | BAD97114.1 | |||
| AK315353.1 | ||||
| BC003620.1 | AAH03620.1 | |||
| BC006202.1 | AAH06202.1 | |||
| BC011869.2 | AAH11869.1 | |||
| BG707314.1 | ||||
| Y17392.1 | CAA76759.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830507 | Chr.5:140156339 - 140334577 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map