Assay Details
Target Gene Details
Entrez Gene ID: | 1456 |
Gene Name: | casein kinase 1 gamma 3 |
Gene Aliases: |
CKI-gamma 3, CSNK1G3L |
Location: |
Chr.5:123512099-123617045 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
More Information
Set Membership: |
|

Genomic Map