Assay Details
Target Gene Details
Entrez Gene ID: | 153643 |
Gene Name: | family with sequence similarity 81 member B |
Gene Aliases: |
- |
Location: |
Chr.5:95384793-95450441 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM81B | NM_152548.2 | NP_689761.2 | ||
| XM_005271905.2 | XP_005271962.2 | |||
| XM_005271906.4 | XP_005271963.2 | |||
| XM_005271907.3 | XP_005271964.1 | |||
| XM_011543207.1 | XP_011541509.1 | |||
| XM_011543208.1 | XP_011541510.1 | |||
| XM_011543209.1 | XP_011541511.1 | |||
| XM_017009118.1 | XP_016864607.1 | |||
| AK058062.1 | BAB71646.1 | |||
| AK310245.1 | ||||
| BC034772.1 | AAH34772.1 | |||
| BM560335.1 |
More Information
Set Membership: |
|

Genomic Map