Assay Details
Target Gene Details
Entrez Gene ID: | 57824 |
Gene Name: | histocompatibility minor HB-1 |
Gene Aliases: |
HB-1, HB-1Y, HLA-HB1 |
Location: |
Chr.5:143812161-143820719 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HMHB1 | NM_021182.1 | NP_067005.1 | ||
| AF103884.1 | AAC78642.1 | |||
| BC104412.1 | ||||
| BC104413.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs77295650] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map