Assay Details
Target Gene Details
Entrez Gene ID: | 84674 |
Gene Name: | caspase recruitment domain family member 6 |
Gene Aliases: |
CINCIN1 |
Location: |
Chr.5:40841190-40855354 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CARD6 | NM_032587.3 | NP_115976.2 | ||
| XM_017009989.1 | XP_016865478.1 | |||
| XM_017009990.1 | XP_016865479.1 | |||
| AB209844.1 | BAD93081.1 | |||
| AF356193.1 | AAK32718.1 | |||
| AK304349.1 | ||||
| AY196783.1 | AAP40637.1 | |||
| BC093825.1 | AAH93825.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv597877 | Chr.5:40796644 - 40848701 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map