Assay Details
Target Gene Details
Entrez Gene ID: | 23367 |
Gene Name: | La ribonucleoprotein domain family member 1 |
Gene Aliases: |
LARP |
Location: |
Chr.5:154682663-154817607 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LARP1 | XM_005268407.3 | XP_005268464.1 | ||
| XM_017009283.1 | XP_016864772.1 | |||
| XM_017009284.1 | XP_016864773.1 |
More Information
Set Membership: |
|

Genomic Map