Assay Details
Target Gene Details
Entrez Gene ID: | 51397 |
Gene Name: | COMM domain containing 10 |
Gene Aliases: |
PTD002 |
Location: |
Chr.5:116084592-116446462 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COMMD10 |
Target Gene Details
Entrez Gene ID: | 101927190 |
Gene Name: | uncharacterized LOC101927190 |
Gene Aliases: |
- |
Location: |
Chr.5:116367416-116374570 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927190 | NR_104675.1 | |||
| BX649078.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1130e214 | Chr.5:116314197 - 116381855 on Build GRCh38 | Loss |
|
| esv2763489 | Chr.5:116357469 - 116397630 on Build GRCh38 | Loss |
|
| esv34146 | Chr.5:115980247 - 116394222 on Build GRCh38 | Loss |
|
| nsv4968 | Chr.5:116300074 - 116379159 on Build GRCh38 | Deletion |
|
| nsv1017983 | Chr.5:116243068 - 116377991 on Build GRCh38 | Loss |
|

Genomic Map