Assay Details
Target Gene Details
Entrez Gene ID: | 54463 |
Gene Name: | family with sequence similarity 134 member B |
Gene Aliases: |
JK-1, JK1 |
Location: |
Chr.5:16473038-16617058 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM134B | NM_001034850.2 | NP_001030022.1 | ||
| NM_019000.4 | NP_061873.2 | |||
| XM_011514053.2 | XP_011512355.1 | |||
| XM_011514054.2 | XP_011512356.1 | |||
| XM_011514055.2 | XP_011512357.1 | |||
| AK024920.1 | BAB15034.1 | |||
| AK025808.1 | BAB15241.1 | |||
| AL832438.1 | CAH10610.1 | |||
| BC030517.1 | AAH30517.1 | |||
| BC053326.1 | AAH53326.1 | |||
| BC073132.1 | AAH73132.1 | |||
| CR994722.1 |
More Information
Set Membership: |
|

Genomic Map