Assay Details
Target Gene Details
Entrez Gene ID: | 202052 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C18 |
Gene Aliases: |
- |
Location: |
Chr.5:139410203-139439525 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC18 | NM_152686.3 | NP_689899.1 | ||
| AK001329.1 | ||||
| AK308138.1 | ||||
| BC030162.2 | AAH30162.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762551 | Chr.5:139396414 - 139447861 on Build GRCh38 | Gain |
|
| nsv519808 | Chr.5:139396890 - 139805924 on Build GRCh38 | Loss |
|

Genomic Map