Assay Details
Target Gene Details
Entrez Gene ID: | 134265 |
Gene Name: | actin filament associated protein 1 like 1 |
Gene Aliases: |
- |
Location: |
Chr.5:149271838-149343637 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AFAP1L1 | NM_001146337.2 | NP_001139809.1 | ||
| NM_001323062.1 | NP_001309991.1 | |||
| NM_001323063.1 | NP_001309992.1 | |||
| NM_152406.3 | NP_689619.1 | |||
| AK074185.1 | ||||
| AK095980.1 | BAC04664.1 | |||
| BC040723.2 | AAH40723.1 | |||
| BC125093.1 | ||||
| BC125094.1 | ||||
| DA792192.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3607138 | Chr.5:149260436 - 149314520 on Build GRCh38 | Gain |
|
| esv3607137 | Chr.5:149260394 - 149353282 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map