Assay Details
Target Gene Details
Entrez Gene ID: | 153768 |
Gene Name: | PRELI domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.5:145587325-145835369 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRELID2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv275206 | Chr.5:145637058 - 145657369 on Build GRCh38 | Loss |
|

Genomic Map