Assay Details
Target Gene Details
Entrez Gene ID: | 411 |
Gene Name: | arylsulfatase B |
Gene Aliases: |
ASB, G4S, MPS6 |
Location: |
Chr.5:78777209-78986534 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARSB | NM_000046.3 | NP_000037.2 | ||
| XM_011543390.1 | XP_011541692.1 | |||
| AK314903.1 | ||||
| J05225.1 | AAA51784.1 | |||
| M32373.1 | AAA51779.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762527 | Chr.5:78625376 - 79252373 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map