Assay Details
Target Gene Details
Entrez Gene ID: | 54826 |
Gene Name: | gypsy retrotransposon integrase 1 |
Gene Aliases: |
GIN-1, TGIN1, ZH2C2 |
Location: |
Chr.5:103086000-103120149 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GIN1 | NM_001317954.1 | NP_001304883.1 | ||
| NM_017676.2 | NP_060146.2 | |||
| NR_134299.1 | ||||
| XM_005272028.3 | XP_005272085.1 | |||
| XM_017009599.1 | XP_016865088.1 | |||
| AK000132.1 | BAA90963.1 | |||
| AK225572.1 | ||||
| AK295796.1 | ||||
| BC015325.1 | AAH15325.1 | |||
| BC151135.1 | ||||
| BC157835.1 | ||||
| CR627424.1 | CAH10511.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830430 | Chr.5:102996203 - 103156228 on Build GRCh38 | Gain |
|
| nsv1029894 | Chr.5:102284375 - 103249595 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74345565] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map