Assay Details
Target Gene Details
Entrez Gene ID: | 3308 |
Gene Name: | heat shock protein family A (Hsp70) member 4 |
Gene Aliases: |
APG-2, HEL-S-5a, HS24/P52, HSPH2, RY, hsp70, hsp70RY |
Location: |
Chr.5:133051970-133105017 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSPA4 | NM_002154.3 | NP_002145.3 | ||
| AB023420.1 | BAA75062.1 | |||
| AB209151.1 | BAD92388.1 | |||
| AK294859.1 | ||||
| AK311097.1 | ||||
| BC002526.2 | AAH02526.1 | |||
| BC110861.1 | AAI10862.1 | |||
| BC126122.1 | ||||
| BC126124.1 | ||||
| BG723797.1 | ||||
| BT007375.1 | AAP36039.1 | |||
| FJ224293.1 | ||||
| L12723.1 | AAA02807.1 | |||
| X67643.1 | CAA47886.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830490 | Chr.5:132953258 - 133115177 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75112461] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map