Assay Details
Target Gene Details
Entrez Gene ID: | 2561 |
Gene Name: | gamma-aminobutyric acid type A receptor beta2 subunit |
Gene Aliases: |
- |
Location: |
Chr.5:161288429-161548124 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GABRB2 | NM_000813.2 | 10 | 4121 | NP_000804.1 |
| NM_021911.2 | 11 | 4235 | NP_068711.1 | |
| AK055125.1 | 1 | 772 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5100 | Chr.5:161264110 - 161308790 on Build GRCh38 | Deletion |
|
| esv3607398 | Chr.5:161183978 - 161302078 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map