Assay Details
Target Gene Details
Entrez Gene ID: | 153643 |
Gene Name: | family with sequence similarity 81 member B |
Gene Aliases: |
- |
Location: |
Chr.5:95384793-95450441 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM81B | XM_005271906.4 | XP_005271963.2 | ||
| XM_017009118.1 | XP_016864607.1 |
More Information
Set Membership: |
|

Genomic Map