Assay Details
Target Gene Details
Entrez Gene ID: | 64097 |
Gene Name: | erythrocyte membrane protein band 4.1 like 4A |
Gene Aliases: |
EPB41L4, NBL4 |
Location: |
Chr.5:112141829-112419490 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPB41L4A | NM_022140.3 | NP_071423.3 | ||
| XM_011543530.1 | XP_011541832.1 | |||
| XM_011543531.2 | XP_011541833.1 | |||
| XM_011543532.1 | XP_011541834.1 | |||
| XM_011543533.1 | XP_011541835.1 | |||
| XM_017009689.1 | XP_016865178.1 | |||
| AA350575.1 | ||||
| AB030240.1 | BAB17229.1 | |||
| BC114942.1 |
Target Gene Details
Entrez Gene ID: | 101927023 |
Gene Name: | uncharacterized LOC101927023 |
Gene Aliases: |
- |
Location: |
Chr.5:112228283-112257309 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927023 | NR_110559.1 | |||
| BU608339.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv599367 | Chr.5:111588583 - 112267016 on Build GRCh38 | Gain |
|
| nsv599371 | Chr.5:112183321 - 112341058 on Build GRCh38 | Loss |
|
| nsv599368 | Chr.5:112064188 - 112696978 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs141352765] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map