Assay Details
Target Gene Details
Entrez Gene ID: | 10814 |
Gene Name: | complexin 2 |
Gene Aliases: |
921-L, CPX-2, CPX2, Hfb1 |
Location: |
Chr.5:175796539-175884021 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPLX2 | NM_006650.3 | NP_006641.1 | ||
| XM_005265799.1 | XP_005265856.1 | |||
| AL707430.1 | ||||
| U35100.1 | AAC50229.1 |
More Information
Set Membership: |
|

Genomic Map