Assay Details
Target Gene Details
Entrez Gene ID: | 4090 |
Gene Name: | SMAD family member 5 |
Gene Aliases: |
DWFC, JV5-1, MADH5 |
Location: |
Chr.5:136132845-136182734 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMAD5 | NM_001001419.2 | NP_001001419.1 | ||
| NM_001001420.2 | NP_001001420.1 | |||
| NM_005903.6 | NP_005894.3 | |||
| XM_017009470.1 | XP_016864959.1 | |||
| AF009745.1 | ||||
| AF071106.1 | AAD20799.1 | |||
| AF071107.1 | AAD20800.1 | |||
| AF071108.1 | AAD20801.1 | |||
| AF071109.1 | AAD20802.1 | |||
| AF071110.1 | AAD20803.1 | |||
| AK311231.1 | ||||
| AK313397.1 | ||||
| BX510058.1 | ||||
| BX646699.1 | ||||
| BX648328.1 | ||||
| CR749366.1 | CAH18219.1 | |||
| CR749473.1 | CAH18303.1 |
More Information
Set Membership: |
|

Genomic Map