Assay Details
Target Gene Details
Entrez Gene ID: | 22936 |
Gene Name: | elongation factor for RNA polymerase II 2 |
Gene Aliases: |
- |
Location: |
Chr.5:95885098-95962288 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELL2 | NM_012081.5 | 12 | 5847 | NP_036213.2 |
| XM_006714575.2 | 13 | 5687 | XP_006714638.1 | |
| XM_017009240.1 | 13 | 6114 | XP_016864729.1 | |
| XM_017009241.1 | 11 | 5942 | XP_016864730.1 | |
| XM_017009242.1 | 12 | 5555 | XP_016864731.1 | |
| XM_017009243.1 | 12 | 6158 | XP_016864732.1 | |
| BX538289.1 | 5 | 4506 | CAD98084.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1015180 | Chr.5:95798653 - 95950742 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map