Assay Details
Target Gene Details
Entrez Gene ID: | 153163 |
Gene Name: | uncharacterized LOC153163 |
Gene Aliases: |
- |
Location: |
Chr.5:122436497-122479087 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MGC32805 | NR_051996.1 |
Target Gene Details
Entrez Gene ID: | 9627 |
Gene Name: | synuclein alpha interacting protein |
Gene Aliases: |
SYPH1, Sph1 |
Location: |
Chr.5:122311354-122464219 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
More Information
Additional Information:
For this assay, SNP(s) [rs148191215] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map