Assay Details
Target Gene Details
Entrez Gene ID: | 84249 |
Gene Name: | pleckstrin and Sec7 domain containing 2 |
Gene Aliases: |
EFA6C |
Location: |
Chr.5:139683980-139844466 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSD2 | XM_017009975.1 | 1 | 1018 | XP_016865464.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830505 | Chr.5:139551597 - 139745300 on Build GRCh38 | Loss |
|
| nsv519808 | Chr.5:139396890 - 139805924 on Build GRCh38 | Loss |
|
| nsv462460 | Chr.5:139553566 - 139765464 on Build GRCh38 | Loss |
|
| nsv830504 | Chr.5:139511905 - 139701120 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map