Assay Details
Target Gene Details
Entrez Gene ID: | 9337 |
Gene Name: | CCR4-NOT transcription complex subunit 8 |
Gene Aliases: |
CAF1, CALIF, Caf1b, POP2, hCAF1 |
Location: |
Chr.5:154857952-154876792 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
More Information
Set Membership: |
|

Genomic Map