Assay Details
Target Gene Details
Entrez Gene ID: | 2566 |
Gene Name: | gamma-aminobutyric acid type A receptor gamma2 subunit |
Gene Aliases: |
CAE2, ECA2, GEFSP3 |
Location: |
Chr.5:162067642-162155539 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GABRG2 | NM_000816.3 | NP_000807.2 | ||
| NM_198903.2 | NP_944493.2 | |||
| NM_198904.2 | NP_944494.1 | |||
| AK299640.1 | ||||
| AK315200.1 | ||||
| BC059389.1 | AAH59389.1 | |||
| BC069348.1 | AAH69348.1 | |||
| BC074795.2 | AAH74795.1 | |||
| CD013896.1 | ||||
| CD013897.1 | ||||
| DA226729.1 | ||||
| X15376.1 | CAA33437.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs114370934] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map